Anti-DC-SIGN/CD209 Mouse Monoclonal Antibody (Biotin) [clone: 111H2.02]
103318-796
- Antibody Type:Primary
- Antigen Symbol:DC-SIGN/CD209
- Clonality:Monoclonal
- Clone:111H2.02
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 μg
- Gene ID:30835
- Antigen Synonyms:CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin|CLEC4LC-type lectin domain family 4 member L|DC-SIGNMGC129965|member L|Dendritic cell-specific ICAM-3-grabbing non-integrin 1|DC-SIGN1C-type lectin domain family 4|CD209 molecule|CDSIGNHIV gpl20-binding protein
- Storage Buffer:Antibody in PBS and 50% Glycerol
- Storage Temperature:Store at -20C in the dark. Avoid freeze-thaw cycles.
- Immunogen:HeLa cells stably transfected-with human DC-SIGN
- Purification:Ion exchange chromatography
- Cat. No.:103318-796
- Supplier no.:DDX0209B
The DC-SIGN / CD209 Antibody (111H2.02) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to DC-SIGN / CD209. This antibody reacts with human. The DC-SIGN / CD209 Antibody (111H2.02) [Biotin] has been validated for the following applications: Immunohistochemistry.
Type: Primary
Antigen: DC-SIGN/CD209
Clonality: Monoclonal
Clone: 111H2.02
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human