Anti-Cx30.1 Rabbit Polyclonal Antibody
103327-206
- Antibody Type:Primary
- Antigen Name:Connexin 30.1
- Antigen Symbol:Cx30.1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:2709
- Antigen Synonyms:connexin 31.1|31.1kDa|connexin-31.1|beta 5|gap junction protein|beta 5 (connexin 31.1)|CX31.1gap junction beta-5 protein|Cx31.1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:GJB5 (NP_005259.1, 1 a.a. - 273 a.a.) full-length human protein. MNWSIFEGLLSGVNKYSTAFGRIWLSLVFIFRVLVYLVTAERVWSDDHKDFDCNTRQPGCSNVCFDEFFPVSHVRLWALQLILVTCPSLLVVMHVAYREVQEKRHREAHGENSGRLYLNPGKKRGGLWWTYVCSLVFKASVDIAFLYVFHSFYPKYILPPVVKCHADPCPNIVDCFISKPSEKNIFTLFMVATAAICILLNLVELIYLVSKRCHECLAARKAQAMCTGHHPHGTTSSCKQDDLLSGDLIFLGSDSHPPLLPDRPRDHVKKTIL
- Purification:Protein G purified
- Cat. No.:103327-206
- Supplier no.:H00002709-D01P
The Connexin 30.1 / GJB5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Connexin 30.1 / GJB5. This antibody reacts with human. The Connexin 30.1 / GJB5 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Cx30.1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human