Anti-CPNE4 Mouse Polyclonal Antibody
Catalog # 103346-798
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CPNE4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Gene ID:131034
- Antigen Synonyms:copine 8|copine-8|CPN4|copine IVCOPN4|MGC15604|copine-4
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CPNE4 (NP_570720.1, 1 a.a. ~ 557 a.a) full-length human protein.MKKMSNIYESAANTLGIFNSPCLTKVELRVACKGISDRDALSKPDPCVILKMQSHGQWFEVDRTEVIRTCINPVYSKLFTVDFYFEEVQRLRFEVHDISSNHNGLKEADFLGGMECTLGQIVSQRKLSKSLLKHGNTAGKSSITVIAEELSGNDDYVELAFNARKLDDKDFFSKSDPFLEIFRMNDDATQQLVHRTEVVMNNLSPAWKSFKVSVNSLCSGDPDRRLKCIVWDWDSNGKHDFIGEFTSTFKEMRGA
- Purification:Protein A purified
- Cat. No.:103346-798
- Supplier no.:H00131034-B01P
Specifications
About this item
The CPNE4 Antibody from Novus Biologicals is a mouse polyclonal antibody to CPNE4. This antibody reacts with human. The CPNE4 Antibody has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: CPNE4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human