Anti-CNOT7 Mouse Monoclonal Antibody [clone: 2F6]
Catalog # 103343-092
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:CNOT7
- Clonality:Monoclonal
- Clone:2F6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 μg
- Gene ID:29883
- Antigen Synonyms:CAF-1|BTG1 binding factor 1|CCR4-NOT transcription complex subunit 7|hCAF-1|BTG1-binding factor 1|CCR4-NOT transcription complex|CAF1carbon catabolite repressor protein (CCR4)-associative factor 1|subunit 7|CCR4-associated factor 1
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:CNOT7 (AAH60852, 1 a.a. ~ 286 a.a) full length recombinant protein with GST tag.MPAATVDHSQRICEVWACNLDEEMKKIRQVIRKYNYVAMDTEFPGVVARPIGEFRSNADYQYQLLRCNVDLLKIIQLGLTFMNEQGEYPPGTSTWQFNFKFNLTEDMYAQDSIELLTTSGIQFKKHEEEGIETQYFAELLMTSGVVLCEGVKWLSFHSGYDFGYLIKILTNSNLPEEELDFFEILRLFFPVIYDVKYLMKSCKNLKGGLQEVAEQLELERIGPQHQAGSDSLLTGMAFFKMREMFFEDHIDDAKY
- Purification:IgG purified
- Cat. No.:103343-092
- Supplier no.:H00029883-M01
Specifications
About this item
The CNOT7 Antibody (2F6) from Novus Biologicals is a mouse monoclonal antibody to CNOT7. This antibody reacts with human, mouse, rat. The CNOT7 Antibody (2F6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: CNOT7
Clonality: Monoclonal
Clone: 2F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG Mix Kappa
Reactivity: Human, Mouse, Rat