To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD39/ENTPD1
- Clonality:Monoclonal
- Clone:AC2.5
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:953
- Antigen Synonyms:DKFZp686I093|NTPDase-1|Ecto-ATP diphosphohydrolase 1|FLJ40959|FLJ40921|Ecto-ATPDase 1|Lymphoid cell activation antigen|Ecto-ATPase 1|Ecto-apyrase|DKFZp686D194|ATPDase|EC 3.6.1|NTPDase 1|CD39EC 3.6.1.5|CD39 antigen|ectonucleoside triphosphate diphosphohydrolase 1
- Storage Buffer:Tris-NaCl (pH 8.0)
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:CD39
- Purification:Ion exchange chromatography
- Cat. No.:103318-842
- Supplier no.:DDX0321P-100
Specifications
About this item
The CD39 / ENTPD1 Antibody (AC2.5) from Novus Biologicals is a mouse monoclonal antibody to CD39 / ENTPD1. This antibody reacts with human. The CD39 / ENTPD1 Antibody (AC2.5) has been validated for the following applications: Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen.
Type: Primary
Antigen: CD39/ENTPD1
Clonality: Monoclonal
Clone: AC2.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human