Anti-Caspase 8 Mouse Monoclonal Antibody (DyLight 755) [clone: 90A992]
Catalog # 103359-720
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Caspase 8
- Clonality:Monoclonal
- Clone:90A992
- Conjugation:DyLight® 755
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Western Blot:Yes
- Size:100 μl
- Gene ID:841
- Antigen Synonyms:AIS|NR3C4KD|spinal and bulbar muscular atrophy|Nuclear receptor subfamily 3 group C member 4|Dihydrotestosterone receptorHYSP1|HUMARA|androgen receptor|DHTRTFM|SMAX1SBMA
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein.
- Purification:Protein G purified
- Cat. No.:103359-720
Specifications
About this item
The Caspase-8 Antibody (90A992) [DyLight 755] from Novus Biologicals is a mouse monoclonal antibody to Caspase-8. This antibody reacts with human, primate. The Caspase-8 Antibody (90A992) [DyLight 755] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Caspase 8
Clonality: Monoclonal
Clone: 90A992
Conjugation: DyLight 755
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primate