To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Caspase 8
- Clonality:Monoclonal
- Clone:90A992
- Conjugation:DyLight® 650
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Primates,Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:841
- Antigen Synonyms:AIS|NR3C4KD|spinal and bulbar muscular atrophy|Nuclear receptor subfamily 3 group C member 4|Dihydrotestosterone receptorHYSP1|HUMARA|androgen receptor|DHTRTFM|SMAX1SBMA
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The immunogen for this product falls between amino acids 360-385 of the human Caspase 8 protein.
- Purification:Protein G purified
- Cat. No.:103359-712
Specifications
About this item
The Caspase-8 Antibody (90A992) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to Caspase-8. This antibody reacts with human, primate. The Caspase-8 Antibody (90A992) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Caspase 8
Clonality: Monoclonal
Clone: 90A992
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Primate