To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Bromodeoxyuridine
- Clonality:Monoclonal
- Clone:29G6.E8
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Antigen Synonyms:BUdr|Bromodeoxyuridine
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Anti-BrdU mononclonal antibody was produced in mice by repeated immunizations prepared via immunizations with BromodeoxyUridine-KLH followed by hybridoma development.
- Purification:Protein A purified
- Cat. No.:103394-352
Specifications
About this item
The Bromodeoxyuridine / BrdU Antibody (29G6.E8) from Novus Biologicals is a mouse monoclonal antibody to Bromodeoxyuridine / BrdU. This antibody reacts with mouse, non-species specific. The Bromodeoxyuridine / BrdU Antibody (29G6.E8) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Bromodeoxyuridine
Clonality: Monoclonal
Clone: 29G6.E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Mouse, Non-species specific