Anti-Avian Flu Matrix Protein 1 Rabbit Polyclonal Antibody
Catalog # 103368-774
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Avian Flu Matrix Protein 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Birds,Chicken
- Western Blot:Yes
- Size:0.025 mg
- Antigen Synonyms:Avian Influenza Matrix Protein 1
- Storage Buffer:PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Amino acids 9-28 of avian flu matrix protein 1 were used as the immunogen.
- Purification:Protein G purified
- Cat. No.:103368-774
Specifications
About this item
The Avian Flu Matrix Protein 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Avian Flu Matrix Protein 1. This antibody reacts with avian, chicken. The Avian Flu Matrix Protein 1 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: Avian Flu Matrix Protein 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Chicken