To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:alpha Tubulin 4a
- Antigen Symbol:TUBA4A
- Clonality:Monoclonal
- Clone:B-5-1-2
- Conjugation:DyLight® 405
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Primates,Chicken,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:7277
- Antigen Synonyms:Tubulin H2-alpha|alpha 4a|alpha 1 (testis specific)|tubulin alpha-4A chain|alpha 1|Tubulin alpha-1 chain|tubulin|FLJ30169|Testis-specific alpha-tubulin|TUBA1|H2-ALPHA|Alpha-tubulin 1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Sarkosyl-resistant filaments from Stronglycentrotus purpuratus (sea urchin).
- Purification:Protein A or G purified
- Cat. No.:103412-946
Specifications
About this item
The alpha Tubulin 4a Antibody (B-5-1-2) [DyLight 405] from Novus Biologicals is a mouse monoclonal antibody to alpha Tubulin 4a. This antibody reacts with human, mouse, rat, bovine, chicken, primate. The alpha Tubulin 4a Antibody (B-5-1-2) [DyLight 405] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: TUBA4A
Clonality: Monoclonal
Clone: B-5-1-2
Conjugation: DyLight 405
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Chicken, Primate