To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:OKT3
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:916
- Antigen Synonyms:CD3G|delta subunit|CD3e molecule|T-cell surface glycoprotein CD3 delta chain|FLJ17620|epsilon (CD3-TCR complex)|CD3g molecule|T3DOKT3|CD3d antigen|CD3 antigen|CD3-GAMMA|delta (CD3-TCR complex)|CD3e antigen|TCRE|FLJ94613|epsilon subunit of T3|MGC138597|CD3d molecule|CD3-DELTA|delta polypeptide (TiT3 complex)|T3E|gamma (CD3-TCR complex)|FLJ18683|FLJ79544|CD3-epsilon|T-cell surface glycoprotein CD3 epsilon chain|CD3g antigen|T-cell receptor T3 delta chain|delta chain|gamma polypeptide (TiT3 complex)|epsilon polypeptide (TiT3 complex)|T-cell antigen receptor complex|FLJ17664|T-cell surface antigen T3/Leu-4 epsilon chain
- Storage Buffer:CD3 antibody at 0.5 mg/ml in PBS containing 0.05% BSA
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Human PBMCs were used as immunogen.
- Purification:Protein G purified
- Cat. No.:103398-410
Specifications
About this item
The CD3 Antibody (OKT3) from Novus Biologicals is a mouse monoclonal antibody to CD3. This antibody reacts with human. The CD3 Antibody (OKT3) has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human