Anti-CD25/IL-2 R alpha Rat Monoclonal Antibody (DyLight 680) [clone: 280414]
103296-354
- Antibody Type:Primary
- Antigen Symbol:CD25/IL-2 R alpha
- Clonality:Monoclonal
- Clone:280414
- Conjugation:DyLight® 680
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:100 μl
- Gene ID:3559
- Antigen Synonyms:interleukin-2 receptor subunit alpha|alpha|TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|interleukin 2 receptor|IDDM10|CD25 antigen|CD25
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant mouse CD25/IL2Ra protein (amino acids 22-236) [UniProt P01590].
- Purification:Protein A or G purified
- Cat. No.:103296-354
- Supplier no.:MAB24383FR
The CD25 / IL-2 R alpha Antibody (280414) [DyLight 680] from Novus Biologicals is a rat monoclonal antibody to CD25 / IL-2 R alpha. This antibody reacts with mouse. The CD25 / IL-2 R alpha Antibody (280414) [DyLight 680] has been validated for the following applications: Flow Cytometry.
Type: Primary
Antigen: CD25/IL-2 R alpha
Clonality: Monoclonal
Clone: 280414
Conjugation: DyLight 680
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse