To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Bromodeoxyuridine
- Antigen Symbol:BrdU
- Clonality:Monoclonal
- Clone:SPM166
- Conjugation:DyLight® 680
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Multiple
- Size:100 μl
- Environmentally Preferable:
- Antigen Synonyms:BUdr|Bromodeoxyuridine
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Bromodeoxyuridine (BrdU) conjugated to KLH
- Purification:Protein G purified
- Cat. No.:103394-474
Specifications
About this item
The Bromodeoxyuridine / BrdU Antibody (SPM166) [DyLight 680] from Novus Biologicals is a mouse monoclonal antibody to Bromodeoxyuridine / BrdU. This antibody reacts with all species. The Bromodeoxyuridine / BrdU Antibody (SPM166) [DyLight 680] has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: SPM166
Conjugation: DyLight 680
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species