Anti-BRD3 Mouse Monoclonal Antibody [clone: 7G6]
Catalog # 103334-584
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:BRD3
- Clonality:Monoclonal
- Clone:7G6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:8019
- Antigen Synonyms:KIAA0043FLJ41328|ORFXbromodomain-containing 3|RING3-like protein|bromodomain-containing protein 3|RING3LFLJ23227|bromodomain containing 3
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:BRD3 (AAH32124 418 a.a. - 556 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EPVEAPALPAPAAPMVSKGAESSRSSEESSSDSGSSDSEEERATRLAELQEQLKAVHEQLAALSQAPVNKPKKKKEKKEKEKKKKDKEKEKEKHKVKAEEEKKAKVAPPAKQAQQKKAPAKKANSTTTAGRDHFLTCGV
- Purification:IgG purified
- Cat. No.:103334-584
- Supplier no.:H00008019-M02
Specifications
About this item
The BRD3 Antibody (7G6) from Novus Biologicals is a mouse monoclonal antibody to BRD3. This antibody reacts with human. The BRD3 Antibody (7G6) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BRD3
Clonality: Monoclonal
Clone: 7G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human