Anti-BRCA1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RAY]
103394-340
- Antibody Type:Primary
- Antigen Symbol:BRCA1
- Clonality:Monoclonal
- Clone:RAY
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Size:100 μl
- Epitope:The epitope recognized is found between amino acids 1314-1600.
- Gene ID:672
- Antigen Synonyms:EC 6.3.2.-|subunit 1|early onset|IRIS|breast cancer type 1 susceptibility protein|PNCA4|breast cancer 1|breast and ovarian cancer susceptibility protein 1|breast and ovarian cancer sususceptibility protein|PSCP|BRCAI|EC 6.3.2
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398]
- Purification:Protein G purified
- Cat. No.:103394-340
The BRCA1 Antibody (RAY) [FITC] from Novus Biologicals is a mouse monoclonal antibody to BRCA1. This antibody reacts with human. The BRCA1 Antibody (RAY) [FITC] has been validated for the following applications: Flow Cytometry, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: RAY
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: The epitope recognized is found between amino acids 1314-1600.
Host: Mouse
Isotype: IgG2a
Reactivity: Human