Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody Type:Primary
- Antigen Symbol:BIN1
- Clonality:Monoclonal
- Clone:12A
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Gene ID:274
- Antigen Synonyms:Amphiphysin-like protein|box dependant MYC interacting protein 1|myc box-dependent-interacting protein 1|Amphiphysin II|AMPH2|SH3P9|MGC10367|bridging integrator 1Box-dependent myc-interacting protein 1|AMPHLDKFZp547F068
- Storage Buffer:0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:12A exon BIN1 protein. Protein accession number NP_004296.1.
- Purification:Protein A purified
- Cat. No.:102713-388
- Supplier no.:NBP2-21689
Specifications
About this item
The BIN1 Antibody (12A) from Novus Biologicals is a mouse monoclonal antibody to BIN1. This antibody reacts with human. The BIN1 Antibody (12A) has been validated for the following applications: Western Blot, ELISA.
Type: Primary
Antigen: BIN1
Clonality: Monoclonal
Clone: 12A
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human