Anti-B7-H2/ICOSLG Rat Monoclonal Antibody [clone: 599829]
103295-568
- Antibody Type:Primary
- Antigen Symbol:B7-H2/ICOSLG
- Clonality:Monoclonal
- Clone:599829
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Reactivity:Mouse
- Size:25ug
- Gene ID:23308
- Antigen Synonyms:ICOS-L|B7-H2B7-related protein 1|B7RP1B7-like protein Gl50|inducible T-cell co-stimulator ligand|CD275|B7RP-1LICOS|GL50transmembrane protein B7-H2 ICOS ligand|CD275 antigen|KIAA0653ICOS ligand|ICOSL|B7H2B7 homolog 2
- Storage Buffer:PBS
- Storage Temperature:Store at -20C in the dark. Avoid freeze-thaw cycles.
- Immunogen:rmB7-H2 aa 47-279
- Purification:Protein A or G purified
- Cat. No.:103295-568
- Supplier no.:MAB1581-SP
The B7-H2 / ICOSLG Antibody (599829) from Novus Biologicals is a rat monoclonal antibody to B7-H2 / ICOSLG. This antibody reacts with mouse. The B7-H2 / ICOSLG Antibody (599829) has been validated for the following applications: Flow Cytometry, CyTOF-ready.
Type: Primary
Antigen: B7-H2/ICOSLG
Clonality: Monoclonal
Clone: 599829
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse