To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ASAH1
- Clonality:Monoclonal
- Clone:1A7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG3 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:427
- Antigen Synonyms:Acylsphingosine deacylase|PHP|acid ceramidase|N-acylsphingosine amidohydrolase|ACDase|Putative 32 kDa heart protein|FLJ21558|ASAH|Acid CDase|N-acylsphingosine amidohydrolase (acid ceramidase) 1|ACEC 3.5.1.23|FLJ22079|PHP32N-acylsphingosine amidohydrolase (acid ceramidase)
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ASAH1 (NP_808592, 25 a.a. - 124 a.a.) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PPWTEDCRKSTYPPSGPTYRGAVPWYTINLDLPPYKRWHELMLDKAPMLKVIVNSLKNMINTFVPSGKVMQVVDEKLPGLLGNFPGPFEEEMKGIAAVTD
- Purification:IgG purified
- Cat. No.:103322-196
- Supplier no.:H00000427-M02
Specifications
About this item
The ASAH1 Antibody (1A7) from Novus Biologicals is a mouse monoclonal antibody to ASAH1. This antibody reacts with human. The ASAH1 Antibody (1A7) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ASAH1
Clonality: Monoclonal
Clone: 1A7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3 Kappa
Reactivity: Human