Anti-ARNT/HIF-1 beta Mouse Monoclonal Antibody (Alexa Fluor® 405) [clone: H1beta234]
Catalog # 103368-008
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:ARNT/HIF-1 beta
- Clonality:Monoclonal
- Clone:H1beta234
- Conjugation:Alexa Fluor® 405
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Primates,Sheep,Ferret,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 μl
- Gene ID:405
- Antigen Synonyms:HIF-1-beta|HIF1BETA|Hypoxia-inducible factor 1-beta|hypoxia-inducible factor 1|HIF-1 Beta|aryl hydrocarbon receptor nuclear translocator|nuclear translocator|ARNT protein|HIF1B|BHLHE2|beta subunit|HIF1-beta
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Fusion protein containing amino acids 496-789 of human HIF-1 beta. [UniProt# P27540]
- Purification:Protein G purified
- Cat. No.:103368-008
Specifications
About this item
The ARNT / HIF-1 beta Antibody (H1beta234) [Alexa Fluor« 405] from Novus Biologicals is a mouse monoclonal antibody to ARNT / HIF-1 beta. This antibody reacts with human, mouse, rat, bovine, ferret, primate, sheep. The ARNT / HIF-1 beta Antibody (H1beta234) [Alexa Fluor« 405] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ARNT/HIF-1 beta
Clonality: Monoclonal
Clone: H1beta234
Conjugation: Alexa Fluor® 405
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, Ferret, Primate, Sheep