Anti-Anthrax LF Mouse Monoclonal Antibody (DyLight 488) [clone: 5F502.2]
103360-580
- Antibody Type:Primary
- Antigen Name:Anthrax Lethal Factor
- Antigen Symbol:Anthrax LF
- Clonality:Monoclonal
- Clone:5F502.2
- Conjugation:DyLight® 488
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Antigen Synonyms:Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|B.anthracis lethal factor|LF|Anthrax LF|bacillus anthracis lethal factor|Lef
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:The antibody was developed by immunizing rabbits with a synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD).
- Purification:Protein G purified
- Cat. No.:103360-580
The Anthrax LF Antibody (5F502.2) [DyLight 488] from Novus Biologicals is a mouse monoclonal antibody to Anthrax LF. This antibody reacts with human. The Anthrax LF Antibody (5F502.2) [DyLight 488] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Anthrax LF
Clonality: Monoclonal
Clone: 5F502.2
Conjugation: DyLight 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human