To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:ANP32A
- Clonality:Monoclonal
- Clone:2G11-4A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:8125
- Antigen Synonyms:PHAPIMGC119787|LANPPutative HLA-DR-associated protein I|PHAP1MGC150373|cerebellar leucine rich acidic nuclear protein|I1PP2A|Potent heat-stable protein phosphatase 2A inhibitor I1PP2A|acidic (leucine-rich) nuclear phosphoprotein 32 family|inhibitor-1 of protein phosphatase-2A|MAPMC15orf1|Mapmodulin|Acidic nuclear phosphoprotein pp32|PP32|putative human HLA class II associated protein I|Leucine-rich acidic nuclear protein|acidic leucine-rich nuclear phosphoprotein 32 family member A|member A
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:ANP32A (AAH07200, 1 a.a. - 249 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEMGRRIHLELRNRTPSDVKELVLDNSRSNEGKLEGLTDEFEELEFLSTINVGLTSIANLPKLNKLKKLELSDNRVSGGLEVLAEKCPNLTHLNLSGNKIKDLSTIEPLKKLENLKSLDLFNCEVTNLNDYRENVFKLLPQLTYLDGYDRDDKEAPDSDAEGYVEGLDDEEEDEDEEEYDEDAQVVEDEEDEDEEEEGEEEDVSGEEEEDEEGYNDGEVDDEEDEEELGEEERGQKRKREPEDEGEDDD
- Purification:IgG purified
- Cat. No.:103334-700
- Supplier no.:H00008125-M01
Specifications
About this item
The ANP32A Antibody (2G11-4A5) from Novus Biologicals is a mouse monoclonal antibody to ANP32A. This antibody reacts with human. The ANP32A Antibody (2G11-4A5) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: ANP32A
Clonality: Monoclonal
Clone: 2G11-4A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human