Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AMPK gamma 1 Mouse Monoclonal Antibody [clone: 7H3E9]
Anti-AMPK gamma 1 Mouse Monoclonal Antibody [clone: 7H3E9]
Catalog # 103413-048
Supplier:  Novus Biologicals
Anti-AMPK gamma 1 Mouse Monoclonal Antibody [clone: 7H3E9]
Catalog # 103413-048
Supplier:  Novus Biologicals
Supplier Number:  NBP2-37285

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AMPK gamma 1
  • Antigen Symbol:
    AMPK gamma 1
  • Clonality:
    Monoclonal
  • Clone:
    7H3E9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 μl
  • Gene ID:
    5571
  • Antigen Synonyms:
    5'-AMP-activated protein kinase subunit gamma-1|gamma-1 subunit|MGC8666|5'-AMP-activated protein kinase|AMPK subunit gamma-1|protein kinase|AMPKg|AMP-activated|gamma 1 non-catalytic subunit|AMPK gamma1|AMPK gamma-1 chain
  • Storage Buffer:
    PBS
  • Storage Temperature:
    Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
  • Concentration:
    1 mg/ml
  • Immunogen:
    Purified recombinant fragment of human PRKAG1 (AA: 230-331) expressed in E. Coli.
  • Purification:
    Protein A or G purified
  • Cat. No.:
    103413-048

Specifications

About this item

The AMPK gamma 1 Antibody (7H3E9) from Novus Biologicals is a mouse monoclonal antibody to AMPK gamma 1. This antibody reacts with human. The AMPK gamma 1 Antibody (7H3E9) has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, CyTOF-ready.

CyTOF-ready

Type: Primary
Antigen: AMPK gamma 1
Clonality: Monoclonal
Clone: 7H3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human