To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:AMPK beta 1
- Antigen Symbol:AMPK beta 1
- Clonality:Monoclonal
- Clone:2C3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 μg
- Environmentally Preferable:
- Gene ID:5564
- Antigen Synonyms:beta-1|AMPK subunit beta-1|noncatalytic|AMPK|HAMPKb|AMPK beta 1,5'-AMP-activated protein kinase subunit beta-1|MGC17785|AMPK beta -1 chain|5'-AMP-activated protein kinase beta-1 subunit|protein kinase|AMP-activated protein kinase beta subunit|AMP-activated|beta 1 non-catalytic subunit|AMPKb
- Storage Buffer:PBS (pH 7.4)
- Storage Temperature:Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
- Immunogen:PRKAB1 (AAH01007, 1 a.a. - 270 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MGNTSSERAALERHGGHKTPRRDSSGGTKDGDRPKILMDSPEDADLFHSEEIKAPEKEEFLAWQHDLEVNDKAPAQARPTVFRWTGGGKEVYLSGSFNNWSKLPLTRSHNNFVAILDLPEGEHQYKFFVDGQWTHDPSEPIVTSQLGTVNNIIQVKKTDFEVFDALMVDSQKCSDVSELSSSPPGPYHQEPYVCKPEERFRAPPILPPHLLQVILNKDTGISCDPALLPEPNHVMLNHLYALSIKDGVMVLSATHRYKKKYVTTLLYKPI
- Purification:IgG purified
- Cat. No.:103308-052
- Supplier no.:H00005564-M02
Specifications
About this item
The AMPK beta 1 Antibody (2C3) from Novus Biologicals is a mouse monoclonal antibody to AMPK beta 1. This antibody reacts with human, mouse. The AMPK beta 1 Antibody (2C3) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: AMPK beta 1
Clonality: Monoclonal
Clone: 2C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse