Anti-AMN1 Rabbit Polyclonal Antibody
Catalog # 103279-256
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Symbol:AMN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μl
- Gene ID:196394
- Antigen Synonyms:protein AMN1 homolog|antagonist of mitotic exit network 1 homolog (S. cerevisiae)
- Storage Buffer:PBS (pH 7.2) and 40% Glycerol
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed against Recombinant Protein corresponding to amino acids:LLHLSNCRKLKKLNLNASKGNRVSVTSEGIKAVASSCSYLHEASLKRCCNLTDEGVVALALNCQLLKIIDLGGCLSITDV
- Purification:Immunogen affinity purified
- Cat. No.:103279-256
- Supplier no.:NBP1-91663
Specifications
About this item
The AMN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to AMN1. This antibody reacts with human. The AMN1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: AMN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human