To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Symbol:AK2
- Clonality:Monoclonal
- Clone:AT7E7
- Conjugation:DyLight® 650
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:204
- Antigen Synonyms:adenylate kinase isoenzyme 2|AK 2|EC 2.7.4.3|EC 2.7.4|adenylate kinase 2|mitochondrial|ADK2|ATP-AMP transphosphorylase 2
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Immunogen:Recombinant human AK2 (1-239aa) purified from E. coli
- Purification:Protein A purified
- Cat. No.:103409-954
The AK2 Antibody (AT7E7) [DyLight 650] from Novus Biologicals is a mouse monoclonal antibody to AK2. This antibody reacts with human. The AK2 Antibody (AT7E7) [DyLight 650] has been validated for the following applications: Western Blot, Flow Cytometry, ELISA, Immunocytochemistry / Immunofluorescence.
Type: Primary
Antigen: AK2
Clonality: Monoclonal
Clone: AT7E7
Conjugation: DyLight 650
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human