Anti-ACVR1 Rabbit Polyclonal Antibody
Catalog # 103359-104
Supplier: Novus Biologicals
Specifications
- Antibody Type:Primary
- Antigen Name:Activin RIA/ALK-2/Activin Receptor Type 1
- Antigen Symbol:ACVR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.025 mg
- Gene ID:90
- Antigen Synonyms:activin A receptor|ACTRI|ALK2|ACVR1A|TSRI|FOP|SKR1|type I|ACVRLK2
- Storage Buffer:PBS containing 0.2% gelatin.
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:A synthetic peptide corresponding to amino acids 475-525 of human ACVR1 was used as immunogen.
- Purification:Protein G purified
- Cat. No.:103359-104
Specifications
About this item
The Activin RIA / ALK-2 / Activin Receptor Type 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Activin RIA / ALK-2 / Activin Receptor Type 1. This antibody reacts with human, mouse, rat. The Activin RIA / ALK-2 / Activin Receptor Type 1 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ACVR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat