To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:17-Beta-Estradiol
- Antigen Symbol:17-Beta-Estradiol
- Clonality:Monoclonal
- Clone:4S24 (BGN/06/8824)
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Multiple
- Size:100 μl
- Antigen Synonyms:17 beta Oestradiol
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Immunogen:17-beta estradiol conjugated to bovine serum albumin at position 6.
- Purification:Protein A or G purified
- Cat. No.:103407-942
The 17-Beta-Estradiol Antibody (4S24 (BGN / 06 / 8824)) [FITC] from Novus Biologicals is a mouse monoclonal antibody to 17-Beta-Estradiol. This antibody reacts with all species. The 17-Beta-Estradiol Antibody (4S24 (BGN / 06 / 8824)) [FITC] has been validated for the following applications: ELISA.
Type: Primary
Antigen: 17-Beta-Estradiol
Clonality: Monoclonal
Clone: 4S24 (BGN/06/8824)
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All Species