ContactUsLinkComponent
Krazy Glue® Maximum Bond Krazy Glue®
Krazy Glue® Maximum Bond Krazy Glue®
Catalog # 102945-060
undefined
Krazy Glue® Maximum Bond Krazy Glue®
Catalog # 102945-060
Supplier Number:  EPIKG48348CO
Specifications
  • Volume:
    0.18 oz.
  • Description:
    Krazy Glue® Maximum Bond Krazy Glue, Extra Strong, Durable, Precision Tip
  • Cat. No.:
    102945-060
  • Supplier no.:
    EPIKG48348MR
Specifications
About this item
Instant-bonding, super-strength glue with no-clog tip.
  • Provides an extra strong, durable, shock-resistant bond
  • Works on ceramic, glass, metal, plastic, wood and rubber
  • Precision tip applicator has an extended, no-clog tip
  • Easy-open, easy to seal hanger cap
  • Larger size and sure-grip panel on the container makes it easier to apply
  • Dries clear
Liquid Super Glue. Clear Color Dried. Clear Color on Application. For adult use.