About this item
This S-Mounting Medium with Acrylic is formulated to permanently adhere the tissue and cover glass to the slide.
- Provides excellent optical quality
- Does not distort or discolor with dyes
- Suitable with both Xylene and Xylene substitutes
S-Mounting Medium with acrylic formula is quick drying and the low viscosity facilitates easy application with no air bubbles. This formula will not yellow or crack over time.