Order Entry
Puerto Rico
ContactUsLinkComponent
Histo-Tek® Mini Linear Slide Stainer, Sakura Finetek
Histo-Tek® Mini Linear Slide Stainer, Sakura Finetek
Catalog # 76176-726
Supplier:  Sakura Finetek
undefined
Histo-Tek® Mini Linear Slide Stainer, Sakura Finetek
Catalog # 76176-726
Supplier:  Sakura Finetek
Supplier Number:  1920
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Histo-Tek® Mini Linear Slide Stainer
  • Dimensions:
    62 W x 22 D x 28 H cm (24.4 x 8.7 x 11")
  • Cat. No.:
    76176-726
  • Electrical:
    100/240 VAC
  • Weight:
    16 kg (35.3 lbs.)

Specifications

About this item

Automated staining for frozen section slides.

  • Simple to use keypad display
  • Continuous loading of up to 4 slides per carrier
  • 50 mL containers to minimize reagent waste and reduce costs
  • Fume cover and filter protects lab personnel

Staining time can be programmed from 2 seconds to 300 seconds for all stations. Program agitation from 0 to 3 dips per station, with intervals of 5 seconds. Start at any station, #1 to #14. Holds up to 16 slides and fits on any benchtop.

Certifications: IECO 61010 classification.