Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Temperature range:Room temperature...75 °C, ±3 °C
- Capacity:66 Slides
- Dimensions:25.5 x 9.75 x 4"
- Cat. No.:75981-540
Specifications
About this item
This sleek, modernized slide warmer is ideal for use in histology labs.
- 66 slide capacity
- Features an LED temperature display
- Anodized black surface provides helpful contrast
This slide warmer is ideal for use in the histology laboratory for slides mounted with tissue sections. The thermal heater ensures that heat is distributed evenly across the surface.