ContactUsLinkComponent
Hematology Stain, Solution D, Fixative, Azer Scientific
Hematology Stain, Solution D, Fixative, Azer Scientific
  10018-834
 :  
Hematology Stain, Solution D, Fixative, Azer Scientific
  10018-834
 :  ES946
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Hematology Stain
  • Size:
    12 x 16 oz.
  • Cat. No.:
    10018-834
  • Supplier no.:
    ES946
Anhydrous ethanol fixative compatible with the Wescor® Aerospray stainer.
Solution D is one of the compatible solutions for the Wescor® Aerospray stainer. Use to achieve superior staining of the eosinophilic and basophilic cell components. This solution should be stored at room temperature.