ContactUsLinkComponent
Active Yeast, Azer Scientific
Active Yeast, Azer Scientific
  75860-348
undefined
Active Yeast, Azer Scientific
  75860-348
  • Description:
    Active Yeast
  • Size:
    2 lbs.
  • Environmentally Preferable:
  • Cat. No.:
    75860-348
Active Yeast for Fly Food
  • Prevents harmful molds from growing in a Drosophila culture
This active yeast is ideal for successful fly culture growth and is often used in conjunction with agar, molasses, and cornmeal.