Cornmeal, Azer Scientific
Cornmeal, 50 lbs.
Catalog # 75860-346
Supplier: VWR International
Jump to:
Product Details & Documents
Cornmeal for Fly Food
- Yellow cornmeal
- Often used with molasses, active yeast, and agar
- For successful fly culture growth
This yellow cornmeal is used for successful fly culture growth. It is often used with molasses, active yeast, and agar.
Specifications
- Description:Cornmeal
- Size:50 lbs.