Ethidium Bromide Greenbag™ Disposal Kit, MP Biomedicals
Catalog # 76020-002
Supplier: MP Biomedicals
CAS Number:
Specifications
- Size:50 Bags
- Cat. No.:76020-002
Specifications
About this item
Each EtBr Green BagTM contains a standard amount of activated material that will safely and easily concentrate Ethidium Bromide from large liquid quantities
One kit has the capacity to remove 500 mg of ethidium bromide from solutions (10 mg EtBr/bag).
The bags containing the biohazardous material can then be disposed of in a proper container.