Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
Catalog # 76195-360
Supplier:  Prosci
Anti-VIM Mouse Monoclonal Antibody [clone: SPM576]
Catalog # 76195-360
Supplier:  Prosci
Supplier Number:  34-182

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    vimentin
  • Antigen Symbol:
    VIM
  • Clonality:
    Monoclonal
  • Clone:
    SPM576
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Goat,
    Chicken,
    Human,
    Bovine,
    Dog,
    Cat
  • Western Blot:
    Yes
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P08670
  • Antigen Synonyms:
    VIM
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Human recombinant protein was used as the immunogen for this VIM antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76195-360

Specifications

About this item

This mAb reacts with a 58kDa protein identified as vimentin. It shows no cross-reaction with other closely related intermediate filament proteins (IFPs) such as desmin, keratin, neurofilament, and glial fibrillary acid protein. Anti-vimentin alone is of limited value as a diagnostic tool; however, when used in panels with other antibodies, it is useful for the sub-classification of a given tumor. Expression of vimentin, when used in conjunction with anti-keratin, is helpful when distinguishing melanomas from undifferentiated carcinomas and large cell lymphomas. All melanomas and Schwannomas react strongly with anti-vimentin. It labels a variety of mesenchymal cells, including melanocytes, lymphocytes, endothelial cells, and fibroblasts. Non-reactivity of anti-vimentin is often considered more useful than its positive reactivity, since there are a few tumors that do not contain vimentin, e.g. hepatoma and seminoma. Anti-vimentin is also useful as a tissue process control reagent.

Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 1-2 ug/ml Western blot: 0.25-0.5 ug/ml Immunohistochemistry (FFPE): 0.25-0.5 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the VIM antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone: SPM576
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Bovine, Dog, Cat, Pig, Goat, Chicken