Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-C4A, C4B Mouse Monoclonal Antibody [clone: SPM545]
Catalog # 76195-346
Supplier:  Prosci
Anti-C4A, C4B Mouse Monoclonal Antibody [clone: SPM545]
Catalog # 76195-346
Supplier:  Prosci
Supplier Number:  34-175

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    complement C4A (Rodgers blood group), complement C4B (Chido blood group)
  • Antigen Symbol:
    C4A, C4B
  • Clonality:
    Monoclonal
  • Clone:
    SPM545
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P0C0L4, P0C0L5
  • Antigen Synonyms:
    C4A, C4B
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Recombinant human C4d protein was used as the immunogen for this Complement 4d antibody.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76195-346

Specifications

About this item

This mAb is specific to Complement 4d (C4d) and it reacts with the secreted as well as cell-bound C4d. C4d is a degradation product of the activated complement factor C4b. Complement 4b is typically activated by binding of Abs to specific target molecules. Following activation and degradation of the C4 molecule, thio-ester groups are exposed, which allow transient, covalent binding of the degradation product Complement 4d to endothelial cell surfaces and extracellular matrix components of vascular basement membranes near the sites of C4 activation. The presence of C4d in peritubular capillaries is a key indicator for acute humoral (i.e. antibody-mediated) rejection of kidney, heart, pancreas and lung allografts. As an established marker of antibody-mediated acute renal allograft rejection and its proclivity for endothelium, this component can be detected in peritubular capillaries in chronic renal allograft rejection as well as hyperacute rejection, acute vascular rejection, acute cellular rejection, and borderline rejection. It has been shown to be a significant predictor of transplant kidney graft survival. Anti-C4d, combined with anti-C3d, can be utilized as a tool for diagnosis of allograft rejection that may warrant a prompt and aggressive anti-rejection treatment.

ELISA: 1-2 ug/ml (order BSA/azide free format) Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the Complement 4d antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: C4A, C4B
Clonality: Monoclonal
Clone: SPM545
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human