Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM528]
Catalog # 76195-256
Supplier:  Prosci
Anti-EPCAM Mouse Monoclonal Antibody [clone: SPM528]
Catalog # 76195-256
Supplier:  Prosci
Supplier Number:  34-130

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    epithelial cell adhesion molecule
  • Antigen Symbol:
    EPCAM
  • Clonality:
    Monoclonal
  • Clone:
    SPM528
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 ug
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    P16422
  • Antigen Synonyms:
    EPCAM
  • Storage Buffer:
    PBS with 0.1 mg/ml BSA and 0.05% sodium azide
  • Storage Temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0.2 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Neuraminidase treated GLS-1 human small cell lung carcinoma cells were used as the immunogen for this EpCAM antibody. The epitope has been localized to amino acids 27-59.
  • Purification:
    Protein G affinity chromatography
  • Cat. No.:
    76195-256

Specifications

About this item

EGP40 is a 40-43kDa transmembrane epithelial glycoprotein, also identified as epithelial specific antigen (ESA), or epithelial cellular adhesion molecule (Ep-CAM). It is expressed on baso-lateral cell surface in most simple epithelia and a vast majority of carcinomas. This antibody has been used to distinguish adenocarcinoma from pleural mesothelioma and hepatocellular carcinoma. This antibody is also useful in distinguishing serous carcinomas of the ovary from mesothelioma.

Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 1-2 ug/ml Western blot: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 min at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the EpCAM antibody for each application should be determined by the researcher.

1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.

Type: Primary
Antigen: EPCAM
Clonality: Monoclonal
Clone: SPM528
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human