Specifications
- Antibody Type:Primary
- Antigen Name:immunoglobulin heavy constant gamma 1 (G1m marker), immunoglobulin heavy constant gamma 2 (G2m marker), immunoglobulin heavy constant gamma 3 (G3m marker), immunoglobulin heavy constant gamma 4 (G4m marker)
- Antigen Symbol:IGHG1, IGHG2, IGHG3, IGHG4
- Clonality:Monoclonal
- Clone:SPM556
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P01857, P01859, P01860, P01861
- Antigen Synonyms:IGHG1, IGHG2, IGHG3, IGHG4
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Purified human Ig Gamma Chain was used as the immunogen for this IgG antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76196-164
Specifications
About this item
Recognizes a protein of 75kDa, identified as gamma heavy chain of human immunoglobulins. It does not cross-react with IgA, IgM, IgE, or IgD, heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This mAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkins lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The optimal dilution of the IgG antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: IGHG1, IGHG2, IGHG3, IGHG4
Clonality: Monoclonal
Clone: SPM556
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human