Specifications
- Antibody Type:Primary
- Antigen Name:extracellular matrix protein 1
- Antigen Symbol:ECM1
- Clonality:Monoclonal
- Clone:SPM217
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q16610
- Antigen Synonyms:ECM1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Secretory Component protein isolated from human colostrum was used as the immunogen for this Secretory Component antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-230
Specifications
About this item
This mAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format: incubate for 30 min at RT (2) The optimal dilution of the Secretory Component antibody for each application should be determined by the researcher.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: SPM217
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human, Rat