Specifications
- Antibody Type:Primary
- Antigen Name:Fli-1 proto-oncogene, ETS transcription factor
- Antigen Symbol:FLI1
- Clonality:Monoclonal
- Clone:FLVI1-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q01543
- Antigen Synonyms:FLI1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human partial recombinant protein (amino acids 107-216) was used as the immunogen for this FLI1 antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76196-124
Specifications
About this item
Friend leukemia integration 1 is a member of the ETS family of DNA binding transcription factors that is involved in cellular proliferation and tumorigenesis. Members of the Ets gene family share a highly conserved carboxy-terminal domain containing a sequence related to the SV40 large T antigen nuclear localization signal sequence. Approximately 90% of Ewing's Sarcoma (EWS) / Primitive Neuroectodermal Tumors (PNET) have a specific translocation, t(11;22)(q24;q12), which results in fusion of EWS to Fli-1, and production of an EWS-Fli-1 fusion protein. Among normal tissues only endothelial cells and small lymphocytes express Fli-1. This protein is expressed in majority of vascular tumors including angiosarcomas, hemangioendotheliomas, hemangiomas, and Kaposi's Sarcomas. High sensitivity and specificity of Fli-1 equals to or exceeds that of the established vascular markers like CD31, CD34, and Factor VIII.
Western blot: 1-2 ug/m;l Flow Cytometry: 0.5-1ug/10^6 cells IF: 1-2 ug/ml The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the FLI1 antibody to be titered up or down for optimal performance.
Type: Primary
Antigen: FLI1
Clonality: Monoclonal
Clone: FLVI1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human