- Antibody Type:Primary
- Antigen Name:enolase 2 (gamma, neuronal)
- Antigen Symbol:ENO2
- Clonality:Monoclonal
- Clone:NSEL1-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P09104
- Antigen Synonyms:ENO2
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Amino acids 416-433 of human Neuron-specific Enolase were used as the immunogen for this NSE antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76196-108
Recognizes a protein of about 50kDa, which is identified as gamma-enolase/neuron specific enolase/enolase 2. Three isoenzymes of enolases are identified, alpha, beta and gamma. Alpha-isoform is expressed in most tissues, whereas beta-form is expressed predominantly in muscle tissue and gamma-enolase is found only in nervous tissue. These isoforms exist as both homodimers and heterodimers, and they play a role in converting phosphoglyceric acid to phosphenolpyruvic acid in the glycolytic pathway. NSE is a useful marker to identify peripheral nerves and tumors of neuro-endocrine origins, such as pheochromocytomas. It it be usually employed in combination with other markers such as Synaptophysin, Chromogranin A, and Neurofilament.
Western blot: 1-2 ug/ml Flow Cytometry: 0.5-1 ug/million cells IF: 1-2 ug/ml IHC (FFPE): 0.5-1 ug/ml (1) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the NSE antibody to be titered up or down for optimal performance.
1. Staining of FFPE tissue requires boiling sections in 10mM Citrate Buffer, pH6, for 10-20 min followed by cooling at RT for 20 min.
Type: Primary
Antigen: ENO2
Clonality: Monoclonal
Clone: NSEL1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human