- Antibody Type:Primary
- Antigen Name:membrane-spanning 4-domains, subfamily A, member 1
- Antigen Symbol:MS4A1
- Clonality:Monoclonal
- Clone:CDLA20-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P11836
- Antigen Synonyms:MS4A1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant CD20 protein was used as the immunogen for this CD20 antibody. The epitope is known to be extracellular.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-952
CD20 is a non-Ig differentiation antigen of B-cells and its expression is restricted to normal and neoplastic B-cells, being absent from all other leukocytes and tissues. It is expressed by pre B-cells and persists during all stages of maturation but is lost upon terminal differentiation into plasma cells. The protein passes through the membrane 4 times with both ends in cytoplasm and exposes one short and one longer loop to the external environment. It acts as a calcium channel involved in B-cell activation and cell cycle progression.
Flow Cytometry: 0.5-1 ug/million cells IF: 0.5-1 ug/ml Immunohistochemistry (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) Titering of the CD20 antibody may be required for optimal performance.
1. FFPE testing requires sections to be boiled in pH6 10mM citrate buffer for 10-20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: CDLA20-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human