- Antibody Type:Primary
- Antigen Name:golgi glycoprotein 1
- Antigen Symbol:GLG1
- Clonality:Monoclonal
- Clone:CRFG1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:Q92896
- Antigen Synonyms:GLG1
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:The Golgi fraction from human liver cells was used as the immunogen for the Golgi antibody. Not suitable for mouse and rat sample testing.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-950
Part of the cellular endomembrane system, the Golgi apparatus packages proteins into membrane-bound vesicles inside the cell before the vesicles are sent to their destination. The Golgi apparatus resides at the intersection of the secretory, lysosomal, and endocytic pathways. It is of particular importance in processing proteins for secretion, containing a set of glycosylation enzymes that attach various sugar monomers to proteins as the proteins move through the apparatus. [Wiki]
This antibody is useful in staining the Golgi apparatus in cell or tissue preparations and subcellular fractions.
Flow Cytometry: 0.5-1 ug/million cells in 0.1ml Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 1-2 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) Titering of the Golgi antibody may be required for optimal performance.
1. FFPE testing requires sections to be boiled in pH6 10mM citrate buffer for 10-20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: GLG1
Clonality: Monoclonal
Clone: CRFG1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human