Specifications
- Antibody Type:Primary
- Antigen Name:small ubiquitin-like modifier 2, small ubiquitin-like modifier 3
- Antigen Symbol:SUMO2, SUMO3
- Clonality:Monoclonal
- Clone:S23MT-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P61956, P55854
- Antigen Synonyms:SUMO2, SUMO3
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human SUMO2 protein was used as the immunogen for this SUMO2/3 antibody. This antibody reacts with both SUMO2 and SUMO3.
- Purification:Protein G purified
- Cat. No.:76194-944
Specifications
About this item
The small ubiquitin-related modifier (SUMO) proteins, which include SUMO1, 2 and 3, belong to the ubiquitin-like protein family. SUMO proteins are synthesized as precursors that undergo processing before conjugation to target proteins. Sumoylation targets for a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. SUMO2/3 contribute to a greater percentage of protein modification than does SUMO1 and they can form polymeric chains. In addition, SUMO3 regulates beta-Amyloid generation and may be critical in the onset or progression of Alzheimer’s disease.
IF: 0.5-1 ug/ml WB: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) Titering of the SUMO2/3 antibody may be required for optimal performance.
1. FFPE testing requires sections to be boiled in pH6 10mM citrate buffer for 10-20 minutes, followed by cooling at RT for 20 minutes, prior to staining.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: SUMO2, SUMO3
Clonality: Monoclonal
Clone: S23MT-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human