Specifications
- Antibody Type:Primary
- Antigen Name:thymidylate synthetase
- Antigen Symbol:TYMS
- Clonality:Monoclonal
- Clone:TYSY9
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P04818
- Antigen Synonyms:TYMS
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Recombinant human protein was used as the immunogen for this Thymidylate synthase antibody.
- Purification:Protein G purified monoclonal antibody
- Cat. No.:76196-076
Specifications
About this item
Thymidylate synthase converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. Thymidylate synthase is also a critical target for the fluoropyrimidines, an important group of anti-neoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates thymidylate synthase by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of thymidylate synthase protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.
Immunofluorescence: 0.5-1 ug/ml IHC (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the Thymidylate synthase antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues is enhanced by boiling tissue sections in 10mM Citrate Buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: TYMS
Clonality: Monoclonal
Clone: TYSY9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human