- Antibody Type:Primary
- Antigen Name:CD8a molecule
- Antigen Symbol:CD8A
- Clonality:Monoclonal
- Clone:CLDA8a-1
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P01732
- Antigen Synonyms:CD8A
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Human recombinant protein was used as the immunogen for this CD8a antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-888
CD8 is a cell surface receptor expressed either as a heterodimer with the beta chain or as a homodimer. A majority of thymocytes and a subpopulation of mature Tcells and NK cells express CD8a. It binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in Tcell development and activation of mature Tcells. For mature Tcells, CD8 and 4 are mutually exclusive, so antibody to CD8 is generally used in conjunction with antibody to CD4. CD8a antibody is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral Tcell lymphomas are CD4+/CD8-. Anaplastic large cell lymphoma is usually CD4+/CD8-, and in T-lymphoblastic lymphoma/leukemia, they are often co-expressed. It is also found in littoral cell angioma of the spleen.
Flow Cytometry: 0.5-1 ug/million cells Immunohistology (FFPE): 0.5-1 ug/ml for 30 minutes at RT (1) Prediluted format : incubate for 30 min at RT (2) The concentration stated for each application is a general starting point. Variations in protocols, secondaries and substrates may require the CD8a antibody to be titered up or down for optimal performance.
1. Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer, pH 6.0, for 10-20 min followed by cooling at RT for 20 minutes.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: CLDA8a-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human