Specifications
- Antibody Type:Primary
- Antigen Name:epidermal growth factor receptor
- Antigen Symbol:EGFR
- Clonality:Monoclonal
- Clone:ERB7-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P00533
- Antigen Synonyms:EGFR
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:Purified EGF receptor was used as the immunogen for the EGFR antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76194-860
Specifications
About this item
Receptor tyrosine kinase binding ligands of the EGF family and activating several signaling cascades to convert extracellular cues into appropriate cellular responses. Known ligands include EGF, TGFA/TGF-alpha, amphiregulin, epigen/EPGN, BTC/betacellulin, epiregulin/EREG and HBEGF/heparin-binding EGF. Ligand binding triggers receptor homo- and/or heterodimerization and autophosphorylation on key cytoplasmic residues. The phosphorylated receptor recruits adapter proteins like GRB2 which in turn activates complex downstream signaling cascades. Activates at least 4 major downstream signaling cascades including the RAS-RAF-MEK-ERK, PI3 kinase-AKT, PLCgamma-PKC and STATs modules. May also activate the NF-kappa-B signaling cascade. Also directly phosphorylates other proteins like RGS16, activating its GTPase activity and probably coupling the EGF receptor signaling to the G protein-coupled receptor signaling. Also phosphorylates MUC1 and increases its interaction with SRC and CTNNB1/beta-catenin. [UniProt]
Immunofluorescence: 1-2 ug/ml Immunohistochemistry (FFPE): 2-4 ug/ml for 30 min at RT (1) Prediluted format : incubate for 30 min at RT (2) Titering of the EGFR antibody may be required for optimal performance.
1. Digest formalin-fixed tissues with Protease at 1mg/ml PBS, pH 7.4 for 10 min at 37oC.
2. The prediluted format is supplied in a dropper bottle and is optimized for use in IHC. After epitope retrieval step (if required), drip mAb solution onto the tissue section and incubate at RT for 30 min.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: ERB7-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, k
Reactivity: Human