Specifications
- Antibody Type:Primary
- Antigen Name:tumor necrosis factor (ligand) superfamily, member 15
- Antigen Symbol:TNFSF15
- Clonality:Monoclonal
- Clone:VEGI/1283
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:O95150
- Antigen Synonyms:TNFSF15
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:A full length human recombinant protein was used as the immunogen for this TL1A antibody.
- Purification:Protein G affinity chromatography
- Cat. No.:76195-966
Specifications
About this item
TNF ligand-related molecule 1 is an anti-angiogenic cytokine that is also called Tumor necrosis factor superfamily, member 15 (TNFSF15) and Vascular endothelial growth inhibitor (VEGI/VEGI192A). This protein is abundantly expressed in endothelial cells, but is not expressed in either B or T cells. The expression of this protein is inducible by TNF and IL-1 alpha. This cytokine is a ligand for receptor TNFRSF25 and decoy receptor TNFRSF21/DR6. It can activate NF-kappaB and MAP kinases, and acts as an autocrine factor to induce apoptosis in endothelial cells. This cytokine is also found to inhibit endothelial cell proliferation, and thus may function as an angiogenesis inhibitor. Reduced expression of TL1A has been reported as a marker of poor prognosis in breast cancer.
Flow Cytometry: 0.5-1ug/10^6 cells IF: 1-2 ug/ml Western blot: 1-2 ug/ml IHC (FFPE): 1-2 ug/ml Titration of the TL1A antibody may be required for optimal performance.
Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: VEGI/1283
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human