Specifications
- Antibody Type:Primary
- Antigen Name:serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
- Antigen Symbol:SERPINA3
- Clonality:Monoclonal
- Clone:AACT/1452
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 ug
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P01011
- Antigen Synonyms:SERPINA3
- Storage Buffer:PBS with 0.1 mg/ml BSA and 0.05% sodium azide
- Storage Temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0.2 mg/mL
- Shipping Temperature:4 °C
- Immunogen:A human partial recombinant protein was used as the immunogen for this Alpha-1-Antichymotrypsin antibody.
- Purification:Protein G affinity
- Cat. No.:76194-446
Specifications
About this item
Alpha 1-Antichymotrypsin, also called AACT and SERPINA3, inhibits the activity of certain proteases, such as cathepsin G that is found in neutrophils, and chymases found in mast cells, by cleaving them into a different shape or conformation. This activity protects some tissues, such as the lower respiratory tract, from damage caused by proteolytic enzymes. This protein is produced in the liver, and is an acute phase protein that is induced during inflammation. [Wiki]
Western blot: 0.5-1 ug/ml IHC (Paraffin): 0.5-1 ug/ml IF: 0.5-1 ug/ml Flow Cytometry: 0.5-1ug/10e6 cells in 0.1ml The stated application concentrations are suggested starting amounts. Titration of the Alpha-1-Antichymotrypsin antibody may be required due to differences in protocols and secondary/substrate sensitivity.
Type: Primary
Antigen: SERPINA3
Clonality: Monoclonal
Clone: AACT/1452
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human